Learn More
R&D Systems™ Human TNF-alpha Quantikine ELISA Kit, R&D Systems™
a 3.5 or 4.5 hour solid phase ELISA designed to measure human TNF-alpha in cell culture supernates, serum, and plasma.
Marke:  R&D Systems™ DTA00C
Weitere Details : Netto-Gewicht : 0.01000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412745
  • 705.24 € / 96 Tests
Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
It contains E. coli-derived recombinant human TNF-alpha and antibodies raised against this protein. It has been shown to accurately quantitate the recombinant factor. Results obtained with naturally occurring TNF-alpha samples showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural human TNF-alpha
Spezifikation
Spezifikation
15.60 - 1000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
7124
1 Plate
5.5pg/mL
Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Natural and recombinant human TNF-alpha
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (200uL), Serum (200uL), EDTA Plasma (200uL), Heparin Plasma (200uL), Citrate Plasma (200uL)
Human
Sufficient materials to run ELISAs on at least one 96-well plate
Videos