Learn More
R&D Systems™ Human Total Bcl-2 DuoSet IC, R&D Systems™
Human Total Bcl-2 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Kreuzreaktivität <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Produkttyp ELISA Development Kit for intracellular proteins
Spezies Human
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously. Complete ELISpot kits are ready-to-r
The Human Total Bcl-2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human Bcl-2.
Spezifikation
Spezifikation
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins