missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total HGFR/c-MET DuoSet IC ELISA
Human Total HGF R/c-MET DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Assay-Bereich | 125.00 to 8000 pg/mL |
---|---|
Kreuzreaktivität | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Gen-ID (Entrez) | 4233 |
Produkttyp | ELISA Development Kit for intracellular proteins |
Konjugat | Biotin |
Beschreibung
The Human Total HGF R/c-MET DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human HGFR/c-MET.Spezifikation
125.00 to 8000 pg/mL | |
4233 | |
Biotin |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit for intracellular proteins | |
Human |