missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total HO-1/HMOX1 DuoSet IC, R&D Systems™
Human Total HO-1/HMOX1 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Assay-Bereich | 156.00 to 10000 pg/mL |
---|---|
Kreuzreaktivität | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Gen-ID (Entrez) | 3162 |
Produkttyp | ELISA Development Kit for intracellular proteins |
Konjugat | Biotin |
Beschreibung
ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. This kit is designed for the detection and enumeration of . Complete ELISpot kits are ready-to-run and require no assay development or refinement.
The Human Total HO-1/HMOX1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human HO-1/HMOX1/HSP32.Spezifikation
156.00 to 10000 pg/mL | |
3162 | |
Biotin |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit for intracellular proteins | |
Human |