missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Total p53 DuoSet IC, R&D Systems™

Human Total p53 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

N.V.

Spezifikation

Assay-Bereich 125.00 to 8000 pg/mL
Kreuzreaktivität <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Gen-ID (Entrez) 7157
Produkttyp ELISA Development Kit for intracellular proteins
Konjugat Biotin
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Human Total p53 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human p53.
Spezifikation

Spezifikation

125.00 to 8000 pg/mL
7157
Biotin
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen