Learn More
Abnova™ Human TSPAN1 Partial ORF (NP_005718, 110 a.a. - 211 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00010103-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. [provided by RefSeq]
Sequence: YTTMAEHFLTLLVVPAIKKDYGSQEDFTQVWNTTMKGLKCCGFTNYTDFEDSPYFKENSAFPPFCCNDNVTNTANETCTKQKAHDQKVEGCFNQLLYDIRTNSpezifikation
NP_005718 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YTTMAEHFLTLLVVPAIKKDYGSQEDFTQVWNTTMKGLKCCGFTNYTDFEDSPYFKENSAFPPFCCNDNVTNTANETCTKQKAHDQKVEGCFNQLLYDIRTN | |
RUO | |
TSPAN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10103 | |
TSPAN1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
9030418M05Rik/NET-1/RP11-322N21.1/TSPAN-1 | |
TSPAN1 | |
Recombinant | |
wheat germ expression system |