missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TXNDC4 Partial ORF (NP_055866.1, 308 a.a. - 406 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00023071-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: LLHIQKTPADCPVIAIDSFRHMYVFGDFKDVLIPGKLKQFVFDLHSGKLHREFHHGPDPTDTAPGEQAQDVASSPPESSFQKLAPSEYRYTLLRDRDELSpezifikation
NP_055866.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ERP44/KIAA0573 | |
TXNDC4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23071 | |
TXNDC4 (Human) Recombinant Protein (Q01) | |
10 ug | |
RUO | |
TXNDC4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |