missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human XPOT Full-length ORF (NP_009166.2, 1 a.a. - 962 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00011260-P01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16166452

  • 482.88 € / 25 Mikrogramm

Voraussichtliches Versanddatum: 13-12-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes a protein belonging to the RAN-GTPase exportin family that mediates export of tRNA from the nucleus to the cytoplasm. Translocation of tRNA to the cytoplasm occurs once exportin has bound both tRNA and GTP-bound RAN. [provided by RefSeq]

Sequence: MDEQALLGLNPNADSDFRQRALAYFEQLKISPDAWQVCAEALAQRTYSDDHVKFFCFQVLEHQVKYKYSELTTVQQQLIRETLISWLQAQMLNPQPEKTFIRNKAAQVFALLFVTEYLTKWPKFFFDILSVVDLNPRGVDLYLRILMAIDSELVDRDVVHTSEEARRNTLIKDTMREQCIPNLVESWYQILQNYQFTNSEVTCQCLEVVGAYVSWIDLSLIANDRFINMLLGHMSIEVLREEACDCLFEVVNKGMDPVDKMKLVESLCQVLQSAGFFSIDQEEDVDFLARFSKLVNGMGQSLIVSWSKLIKNGDIKNAQEALQAIETKVALMLQLLIHEDDDISSNIIGFCYDYLHILKQLTVLSDQQKANVEAIMLAVMKKLTYDEEYNFENEGEDEAMFVEYRKQLKLLLDRLAQVSPELLLASVRRVFSSTLQNWQTTRFMEVEVAIRLLYMLAEALPVSHGAHFSGDVSKASALQDMMRTLVTSGVSSYQHTSVTLEFFETVVRYEKFFTVEPQHIPCVLMAFLDHRGLRHSSAKVRSRTAYLFSRFVKSLNKQMNPFIEDILNRIQDLLELSPPENGHQSLLSSDDQLFIYETAGVLIVNSEYPAERKQALMRNLLTPLMEKFKILLEKLMLAQDEERQASLADCLNHAVGFASRTSKAFSNKQTVKQCGCSEVYLDCLQTFLPALSCPLQKDILRSGVRTFLHRMIICLEEEVLPFIPSASEHMLKDCEAKDLQEFIPLINQITAKFKIQVSPFLQQMFMPLLHAIFEVLLRPAEENDQSAALEKQMLRRSYFAFLQTVTGSGMSEVIANQGAENVERVLVTVIQGAVEYPDPIAQKTCFIILSKLVELWGGKDGPVGFADFVYKHIVPACFLAPLKQTFDLADAQTVLALSECAVTLKTIHLKRGPECVQYLQQEYLPSLQVAPEIIQEFCQALQQPDAKVFKNYLKVFFQRAKP
Spezifikation

Spezifikation

NP_009166.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
136.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
XPO3
XPOT
Yes
wheat germ expression system
Liquid
11260
XPOT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDEQALLGLNPNADSDFRQRALAYFEQLKISPDAWQVCAEALAQRTYSDDHVKFFCFQVLEHQVKYKYSELTTVQQQLIRETLISWLQAQMLNPQPEKTFIRNKAAQVFALLFVTEYLTKWPKFFFDILSVVDLNPRGVDLYLRILMAIDSELVDRDVVHTSEEARRNTLIKDTMREQCIPNLVESWYQILQNYQFTNSEVTCQCLEVVGAYVSWIDLSLIANDRFINMLLGHMSIEVLREEACDCLFEVVNKGMDPVDKMKLVESLCQVLQSAGFFSIDQEEDVDFLARFSKLVNGMGQSLIVSWSKLIKNGDIKNAQEALQAIETKVALMLQLLIHEDDDISSNIIGFCYDYLHILKQLTVLSDQQKANVEAIMLAVMKKLTYDEEYNFENEGEDEAMFVEYRKQLKLLLDRLAQVSPELLLASVRRVFSSTLQNWQTTRFMEVEVAIRLLYMLAEALPVSHGAHFSGDVSKASALQDMMRTLVTSGVSSYQHTSVTLEFFETVVRYEKFFTVEPQHIPCVLMAFLDHRGLRHSSAKVRSRTAYLFSRFVKSLNKQMNPFIEDILNRIQDLLELSPPENGHQSLLSSDDQLFIYETAGVLIVNSEYPAERKQALMRNLLTPLMEKFKILLEKLMLAQDEERQASLADCLNHAVGFASRTSKAFSNKQTVKQCGCSEVYLDCLQTFLPALSCPLQKDILRSGVRTFLHRMIICLEEEVLPFIPSASEHMLKDCEAKDLQEFIPLINQITAKFKIQVSPFLQQMFMPLLHAIFEVLLRPAEENDQSAALEKQMLRRSYFAFLQTVTGSGMSEVIANQGAENVERVLVTVIQGAVEYPDPIAQKTCFIILSKLVELWGGKDGPVGFADFVYKHIVPACFLAPLKQTFDLADAQTVLALSECAVTLKTIHLKRGPECVQYLQQEYLPSLQVAPEIIQEFCQALQQPDAKVFKNYLKVFFQRAKP
RUO
XPOT
Wheat Germ (in vitro)
GST
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate