missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZIC3 Partial ORF (NP_003404.1, 182 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00007547-Q01.2ug

Weitere Details : Netto-Gewicht : 0.00010kg

Artikelnummer. 16197675

  • 229.00 € / 2 Mikrogramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs. [provided by RefSeq]

Sequence: NQVHLGLRGELFGRADPYRPVASPRTDPYAAGAQFPNYSPMNMNMGVNVAAHHGPGAFFRYMRQPIKQELSCKWIDEAQLSRPKKSCDRTFST
Spezifikation

Spezifikation

NP_003404.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.86kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HTX/HTX1/ZNF203
ZIC3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7547
ZIC3 (Human) Recombinant Protein (Q01)
12.5% SDS-PAGE Stained with Coomassie Blue.
NQVHLGLRGELFGRADPYRPVASPRTDPYAAGAQFPNYSPMNMNMGVNVAAHHGPGAFFRYMRQPIKQELSCKWIDEAQLSRPKKSCDRTFST
RUO
ZIC3
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote