Learn More
Abnova™ Human ZNF181 Partial ORF (XP_290835, 354 a.a. - 461 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00339318-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins. See MIM 604749 for additional information on zinc finger proteins.[supplied by OMIM]
Sequence: FIHRSSLIHHQKIHTGEKPYECRECGKAFCCSSHLTRHQRIHTMEKQYECNKCLKVFSSLSFLVQHQSIHTEEKPFECQKCRKSFNQLESLNMHLRNHIRLKPYECSISpezifikation
XP_290835 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FIHRSSLIHHQKIHTGEKPYECRECGKAFCCSSHLTRHQRIHTMEKQYECNKCLKVFSSLSFLVQHQSIHTEEKPFECQKCRKSFNQLESLNMHLRNHIRLKPYECSI | |
RUO | |
ZNF181 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
339318 | |
ZNF181 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HHZ181/MGC44316 | |
ZNF181 | |
Recombinant | |
wheat germ expression system |