missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF32 Full-length ORF (NP_001005368.1, 1 a.a. - 273 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00007580-P01.2ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: MFGFPTATLLDCHGRYAQNVAFFNVMTEAHHKYDHSEATGSSSWDIQNSFRREKLEQKSPDSKTLQEDSPGVRQRVYECQECGKSFRQKGSLTLHERIHTGQKPFECTHCGKSFRAKGNLVTHQRIHTGEKPYQCKECGKSFSQRGSLAVHERLHTGQKPYECAICQRSFRNQSNLAVHRRVHSGEKPYRCDQCGKAFSQKGSLIVHIRVHTGLKPYACTQCRKSFHTRGNCILHGKIHTGETPYLCGQCGKSFTQRGSLAVHQRSCSQRLTLSpezifikation
NP_001005368.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KOX30 | |
ZNF32 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7580 | |
ZNF32 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MFGFPTATLLDCHGRYAQNVAFFNVMTEAHHKYDHSEATGSSSWDIQNSFRREKLEQKSPDSKTLQEDSPGVRQRVYECQECGKSFRQKGSLTLHERIHTGQKPFECTHCGKSFRAKGNLVTHQRIHTGEKPYQCKECGKSFSQRGSLAVHERLHTGQKPYECAICQRSFRNQSNLAVHRRVHSGEKPYRCDQCGKAFSQKGSLIVHIRVHTGLKPYACTQCRKSFHTRGNCILHGKIHTGETPYLCGQCGKSFTQRGSLAVHQRSCSQRLTL | |
RUO | |
ZNF32 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |