missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF75 Partial ORF (NP_009062.2, 249 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00007626-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: WQLLNPLEKTLYNDVMQDIYETVISLGLKLKNDTGNDHPISVSTSEIQTSGCEVSKKTRMKIAQKTMGRENPGDTHSVQKWHRSpezifikation
NP_009062.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.87kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
WQLLNPLEKTLYNDVMQDIYETVISLGLKLKNDTGNDHPISVSTSEIQTSGCEVSKKTRMKIAQKTMGRENPGDTHSVQKWHR | |
RUO | |
ZNF75D | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7626 | |
ZNF75 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D8C6/MGC126327/ZNF75/ZNF82 | |
ZNF75D | |
Recombinant | |
wheat germ expression system |