missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ HumanKine™ Recombinant Human SCF Protein

Artikelnummer. 16811286 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
10 μg
100 μg
1000 μg
Packungsgröße:
10 Mikrogramm
100 Mikrogramm
1000 Mikrogramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16811286 100 μg 100 Mikrogramm
16821286 10 μg 10 Mikrogramm
16801286 1000 μg 1000 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16811286 Lieferant Proteintech™ Lieferanten-Nr. HZ1024100UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells

  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity

Spezifikation

Gen-ID (Entrez) 4254
Molekulargewicht 35 to 45 kDa, monomer, glycosylated
Menge 100 μg
Spezies Human
Rekonstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Inhalt und Lagerung Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Aktivität Typically ≤ 25 ng/mL EC50
Expressionssystem Human embryonic kidney 293 (HEK293) human cell expression system
Form Lyophilized
Reinheits- oder Qualitätsgrad > 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.