Learn More
Huntingtin Recombinant Rabbit Monoclonal Antibody (3H2L4), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen 701663
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with Monkey, Mouse and Bovine. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Huntingtin Recombinant Monoclonal antibody specifically detects Huntingtin in Human samples. It is validated for Immunocytochemistry.Spezifikation
Huntingtin | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P42858 | |
HTT | |
Peptides corresponding to Human HTT (aa 584-601, 650-666, 82-99) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry | |
3H2L4 | |
Unconjugated | |
HTT | |
AI256365; C430023I11Rik; Hd; HD protein; HD protein homolog; Hdh; HTT; Huntingtin; huntingtin (Huntington disease); Huntingtin, myristoylated N-terminal fragment; Huntington disease gene homolog; huntington disease protein; huntington disease protein homolog; IT15; solute carrier family 6 (neurotransmitter transporter, serotonin), member 4 | |
Rabbit | |
Protein A | |
RUO | |
3064 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |