missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYAL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550426
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total HYAL2 protein.
HYAL2 Polyclonal antibody specifically detects HYAL2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
HYAL2 | |
Polyclonal | |
Unconjugated | |
HYAL2 | |
AI256841; AU020858; AW555733; HYAL2; Hyal-2; Hyaluronidase; hyaluronidase 2; hyaluronidase-2; hyaluronoglucosaminidase 2; hyaluronoglucosaminidase-2; LUCA2; LuCa-2; lung carcinoma protein 2; lysosomal hyaluronidase; PH20 homolog; PH-20 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8692 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q12891 | |
HYAL2 | |
Fusion protein corresponding to a region derived from internal residues of human hyaluronoglucosaminidase 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |