Learn More
Hygromycin B, BLD Pharm™
Product Code. 30966900
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30966900 250 mg 250mg
30952232 1 g 1g
30956659 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30966900 Supplier BLD Pharm Supplier No. BD134243250mg
 Restricted Item
Verfügbar ab: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
≥950 μ/mg
Specifications
CAS 31282-04-9
Molecular Formula C20H37N3O13
MDL Number MFCD06795479
Quantity 250 mg
SMILES O[C@H]([C@H]1O)C2(O[C@@]([C@@H](N)CO)([H])[C@@H]1O)O[C@@]([C@@H](O[C@@H]3CO)O[C@@]([C@H]([C@H](N)C4)O)([H])[C@@H]([C@H]4NC)O)([H])[C@@]([C@H]3O)([H])O2
Molecular Weight (g/mol) 527.52
Chemical Name or Material Hygromycin B