missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17170-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The Trip230 gene encodes a protein of 1,978 amino acids with an estimated molecular mass of 230 kDa. The gene is located on chromosome 14q31, which is in a region associated with genetic alterations and thyroid hormone response abnormalities. The Trip230 protein interacts with Rb and thyroid hormone receptor (TR) using two distinct domains. Complex formation with Rb is in a hormone independent manner, but when Trip230 forms a complex with TR, it is in a thyroid hormone-dependent manner. After T3 treatment, Trip230 is phosphorylated and transported to the nucleus, suggesting that phosphorylation may be important for its nuclear translocation.Spezifikation
IARS2 | |
Polyclonal | |
Unconjugated | |
IARS2 | |
IARS2, IleRS, Isoleucine tRNA ligase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
364070, 381314, 55699 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BIJ6, Q9NSE4 | |
Iars2 | |
IARS2 Fusion Protein Ag10696 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |