missing translation for 'onlineSavingsMsg'
Learn More

Ibutilid-Hemifumarat-Salz, 99 %, Thermo Scientific Chemicals

Catalog Number p-7037587
Click to view available options
Menge:
25 mg
100 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
122647-32-9
C44H76N4O10S2
885.23
MFCD01715410
PCIOHQNIRPWFMV-WXXKFALUNA-N
ibutilide fumarate, corvert, ibutilide fumarate usan, ibutilide hemifumarate, ibutilide hemifumarate salt, corvert tn, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfoanilide e-2-butenedioate 2:1, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfonanilide fumarate 2:1 salt, +--n-4-4-ethylheptylamino-1-hydroxybutyl phenyl methanesulfonamide e-butenedioate
5281065
(2E)-but-2-enedioic acid; bis(N-(4-{4-[ethyl(heptyl)amino]-1-hydroxybutyl}phenyl)methanesulfonamide)
OC(=O)\C=C\C(O)=O.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
122647-32-9
C44H76N4O10S2
885.23
MFCD01715410
PCIOHQNIRPWFMV-WXXKFALUNA-N
ibutilide fumarate, corvert, ibutilide fumarate usan, ibutilide hemifumarate, ibutilide hemifumarate salt, corvert tn, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfoanilide e-2-butenedioate 2:1, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfonanilide fumarate 2:1 salt, +--n-4-4-ethylheptylamino-1-hydroxybutyl phenyl methanesulfonamide e-butenedioate
5281065
(2E)-but-2-enedioic acid; bis(N-(4-{4-[ethyl(heptyl)amino]-1-hydroxybutyl}phenyl)methanesulfonamide)
OC(=O)\C=C\C(O)=O.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ibutilide hemifumarate salt
Menge 25 mg
Merck Index 14,4883
Formelmasse 442.61
Reinheit (%) 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'