missing translation for 'onlineSavingsMsg'
Learn More

ICAM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10121-2-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16886293

  • 112.71 € / 20 Mikroliter

Verfügbar ab
16-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody got 55-80 kDa in western blotting maybe due to glycosylation.

ICAM proteins are ligands for the leukocyte adhesion protein LFA-1 (integrin alpha-L/beta-2). ICAM2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance.
Spezifikation

Spezifikation

ICAM2
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P13598, P35330
CD102, ICAM 2, ICAM2
Rabbit
IgG
20 μL
-20°C
Primary
15896, 3384, 360647
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
ICAM2
Liquid
ICAM2
ICAM2 Fusion Protein Ag0165
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation