Learn More
Sino Biological™ ICAM3 Recombinant Human Protein
Expressed from DNA sequence encoding extracellular domain of human ICAM3 (Met 1-His 485) fused to C-terminal fused polyhistidine tag
Marke: Sino Biological™ 10333H08H50
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Expressed from DNA sequence encoding extracellular domain of human ICAM3 (Met 1-His 485) fused to C-terminal fused polyhistidine tag
- Measured by ability of immobilized protein to support PMA adhesion stimulated HSB2 human peripheral blood acute lymphoblastic leukemia cells
- When cells are added to ICAM3 coated plates (12.5™g/mL , 100μL/well), approximately 5% -1
Spezifikation
Protein Structure Analysis, Assay Standard | |
50.8kd | |
Human | |
Truncated, Recombinant | |
Intercellular Adhesion Molecule 3 | |
ICAM3 Recombinant Human Protein | |
Mammalian | |
≥90% by SDS-PAGE |
3385 | |
50μg | |
−20°C | |
<1 EU/μg | |
ICAM3 | |
His Tag | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.