Learn More
IDO Monoclonal Antibody (eyedio), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
131.00 € - 378.00 €
Spezifikation
Antigen | IDO |
---|---|
Klon | eyedio |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16340934
|
Affymetrix eBioscience
48-9477-42 |
100 Tests |
378.00 €
100 Stück |
Verfügbar ab: 13-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16330934
|
Affymetrix eBioscience
48-9477-41 |
25 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: This eyedio monoclonal antibody reacts with human indoleamine-2,3-dioxygenase (IDO, INDO, IDO1), an intracellular enzyme that catalyzes the degradation of tryptophan to kynurenines. IDO is expressed in a wide variety of tissues and cells, including macrophages, plasmacytoid dendritic cells, and several cell lines. IDO can be induced in many different cell types by IFN gamma or other inflammatory stimuli. Expression of IDO in antigen presenting cells and tumors is thought to mediate immune suppression through depletion of this essential amino acid and/or by the creation of tryptophan metabolites that cause apoptosis of T cells and induction of regulatory T cells. Applications Reported: This eyedio antibody has been reported for use in flow cytometric analysis. Applications Tested: This eyedio antibody has been pre-diluted and tested by flow cytometric analysis of stimulated mouse splenocytes using the Intracellular Fixation & Permeabilization Buffer Set (Product No. 88-8824-00) and protocol. Please refer to Staining Intracellular Antigens for Flow Cytometry, Protocol A: Two step protocol for intracellular (cytoplasmic) proteins located on the Resources Tab online. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
IDO Monoclonal antibody specifically detects IDO in Human samples. It is validated for Flow Cytometry.Spezifikation
IDO | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P14902 | |
IgG1 κ | |
4° C, store in dark, DO NOT FREEZE! |
eyedio | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human | |
3620 | |
Primary | |
Antibody |