missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Iduronate 2-Sulfatase/IDS Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
N.V.
Spezifikation
Antigen | Iduronate 2-Sulfatase/IDS |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Anwendungen | Western Blot, Immunoprecipitation |
Klassifikation | Polyclonal |
Beschreibung
Iduronate 2-Sulfatase/IDS Polyclonal specifically detects Iduronate 2-Sulfatase/IDS in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Spezifikation
Iduronate 2-Sulfatase/IDS | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
Alpha-L-iduronate sulfate sulfatase, EC 3.1.6.13, iduronate 2-sulfatase, iduronate 2-sulfatase 14 kDa chain, iduronate 2-sulfatase 42 kDa chain, idursulfase, MPS2, S, SIDS | |
IDS | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
Q08890 | |
3423 | |
Mouse myeloma cell line NS0-derived recombinant mouse Iduronate 2-Sulfatase/IDS Thr36-Pro552 Accession # Q08890 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Iduronate 2-Sulfatase/IDS in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human IDS is observed. |
For Research Use Only