missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IFI27 (Human) Recombinant Protein
Human IFI27 full-length ORF ( AAH15492, 1 a.a. - 119 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00003429-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MEASALTSSAVTSVAKVVRVASGSAVVLPLARIATVVIGGVVAVPMVLSAMGFTAAGIASSSIAAKMMSAAAIANGGGVASGSLVATLQSLGATGLSGLTKFILGSIGSAIAAVIARFY
Spezifikation
AAH15492 | |
interferon, alpha-inducible protein 27 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IFI27 | |
GST |
3429 | |
Wheat germ expression system | |
25 μg | |
FAM14D, ISG12, ISG12A, P27 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |