Learn More
IgE Monoclonal Antibody (Ige21), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
141.00 € - 312.00 €
Spezifikation
Antigen | IgE |
---|---|
Klon | Ige21 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Super Bright 436 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16341314
|
Affymetrix eBioscience
62-6986-42 |
100 Tests |
312.00 €
100 Stück |
Verfügbar ab: 14-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16331314
|
Affymetrix eBioscience
62-6986-41 |
25 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: The monoclonal antibody IgE21 recognizes the immunoglobulin E class of antibodies. The natural levels of IgE in the serum are quite low, comprising only a small portion of the total immunoglobulin. IgE plays a role in allergy responses by binding to mast cells and basophils through the Fc epsilon receptor I which results in the release of histamine and other molecules. The IgE is produced by terminally differentiated plasma B cells and basophils. Like all immunoglobulins, IgE is expressed as a soluble molecule as well as a membrane bound form. IgE21 recognizes both forms. Applications Reported: This IgE21 antibody has been reported for use in flow cytometric analysis. Applications Tested: This IgE21 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No.
IgE Monoclonal antibody specifically detects IgE in Human samples. It is validated for Flow Cytometry.Spezifikation
IgE | |
Flow Cytometry | |
Super Bright 436 | |
Mouse | |
Human | |
3497 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
Ige21 | |
Monoclonal | |
Liquid | |
RUO | |
P01854 | |
IGHE | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IGHE |