missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Fraction to beta-Galactosidase, Unlabeled, Polyclonal, MP Biomedicals
Goat Polyclonal Antibody
Marke: MP Biomedicals 0856028
Weitere Details : Netto-Gewicht : 0.00100kg
Beschreibung
beta-Galactosidase Polyclonal antibody specifically detects beta-Galactosidase in samples. It is validated for Enzyme Immunoassay, Fluorescent Immunoassay, Immunohistochemistry, Western Blotting.Spezifikation
IgG Fraction to beta-Galactosidase | |
Polyclonal | |
Goat | |
1 mL | |
Store unopened product at 2° to 8°C. For long term storage, aliquot and freeze at -20°C. Avoid repeated freeze/thaw cycles. |
ELISA, Fluorescent Immunoassay (FIA), Immunohistochemistry, Western Blot | |
Unconjugated | |
Recombinant full length protein of Escherichia coli beta-Galactosidase | |
Primary |