Learn More
IgM Monoclonal Antibody (SA-DA4), FITC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
93.25 € - 210.00 €
Spezifikation
Antigen | IgM |
---|---|
Klon | SA-DA4 |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry, Immunohistochemistry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15556666
|
Affymetrix eBioscience
11-9998-41 |
25 Tests |
93.25 €
25 Tests |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen | Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15566666
|
Affymetrix eBioscience
11-9998-42 |
100 Tests |
210.00 €
100 Tests |
Verfügbar ab: 02-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Description: The SA-DA4 monoclonal antibody reacts with the mu heavy chain of human IgM. It does not react with other classes of human immunoglobulin including IgD, IgG, or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of mature B cells. Our testing has revealed variations in staining of the non-B cell population that is predicted to be non-specific. This variation can be attributed to both donor variability and the anticoagulant used during blood draw. Therefore, we recommend using sodium citrate as the anticoagulant to minimize non-specific staining by this antibody. Applications Reported: This SA-DA4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SA-DA4 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.Spezifikation
IgM | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH | |
IGHM | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IGHM |
SA-DA4 | |
Flow Cytometry, Immunohistochemistry | |
FITC | |
Mouse | |
Human | |
P01871 | |
3507 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.