Learn More
IL-1 beta Monoclonal Antibody (CRM56), Alexa Fluor™ 647, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
108.00 € - 266.00 €
Spezifikation
Antigen | IL-1 beta |
---|---|
Klon | CRM56 |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16320964
|
Affymetrix eBioscience
51-7018-42 |
100 Tests |
266.00 €
100 Stück |
Verfügbar ab: 13-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16310964
|
Invitrogen
51-7018-41 |
25 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: The CRM56 antibody reacts with human and baboon interleukin-1 beta; (IL-1 beta). IL-1 beta is a 17 kDa factor secreted primarily by monocytes. IL-1 has effects on T-helper cells, inducing IL-2 secretion and expression of IL-2 receptors. IL-1 has effects on B cells, promoting cell proliferation and immunoglobulin synthesis. Applications Reported: This CRM56 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CRM56 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to "e;Staining Intracellular Antigens for Flow Cytometry, Protocol A: Two step protocol for intracellular (cytoplasmic) proteins."e; This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser
IL-1 beta Monoclonal antibody specifically detects IL-1 beta in Human samples. It is validated for Flow Cytometry.Spezifikation
IL-1 beta | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P01584 | |
IL1B | |
Primary | |
Antibody |
CRM56 | |
Flow Cytometry | |
Alexa Fluor 647 | |
Mouse | |
Human | |
3553 | |
IgG1 | |
4° C, store in dark, DO NOT FREEZE! | |
IL1B |