Learn More
IL-13 Monoclonal Antibody (85BRD), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
126.00 € - 354.00 €
Spezifikation
Antigen | IL-13 |
---|---|
Klon | 85BRD |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16320934
|
Affymetrix eBioscience
48-7136-42 |
100 Tests |
354.00 €
100 Stück |
Verfügbar ab: 17-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16310934
|
Affymetrix eBioscience
48-7136-41 |
25 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: The 85BRD monoclonal antibody reacts with human interleukin-13 (IL-13). IL-13, also known as NC30, is a 12.5 kDa protein secreted by activated T helper cells, CD8+ T cells, and NK cells. The biological activities of IL-13 include suppression of macrophage cytotoxic activity, upregulation of IL-1RA expression and suppression of inflammatory cytokine expression. Applications Reported: This 85BRD antibody has been reported for use in flow cytometric analysis. Applications Tested: This 85BRD antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to "e;Staining Intracellular Antigens for Flow Cytometry, Protocol A: Two step protocol for intracellular (cytoplasmic) proteins."e; This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor 450 is an alternative to Pacific Blue. eFluor 450 emits at 446 nm and is excited with the violet laser line (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser
IL-13 Monoclonal antibody specifically detects IL-13 in Human samples. It is validated for Flow Cytometry.Spezifikation
IL-13 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P35225 | |
IgG2b κ | |
4° C, store in dark, DO NOT FREEZE! |
85BRD | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human | |
3596 | |
Primary | |
Antibody |