missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-19 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24402-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a cytokine that belongs to the IL10 cytokine subfamily. This cytokine is found to be preferentially expressed in monocytes. It can bind the IL20 receptor complex and lead to the activation of the signal transducer and activator of transcription 3 (STAT3). A similar cytokine in mouse is reported to up-regulate the expression of IL6 and TNF-alpha and induce apoptosis, which suggests a role of this cytokine in inflammatory responses. Alternatively spliced transcript variants encoding the distinct isoforms have been described.Spezifikation
IL-19 | |
Polyclonal | |
Unconjugated | |
IL19 | |
IL 10C, IL 19, IL19, interleukin 19, MDA1, NG.1, ZMDA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29949 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UHD0 | |
IL19 | |
IL-19 Fusion Protein Ag21496 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |