missing translation for 'onlineSavingsMsg'
Learn More

IL-2 Monoclonal Antibody (MQ1-17H12), Alexa Fluor™ 532, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

181.00 € - 443.00 €

Spezifikation

Antigen IL-2
Klon MQ1-17H12
Konzentration 5 μl/Test
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16321064
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
58-7029-41
25 Tests
181.00 €
25 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
16331064
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
58-7029-42
100 Tests
443.00 €
100 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung

Beschreibung

Description: The MQ1-17H12 antibody reacts with human interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine. The MQ1-17H12 antibody is a non-neutralizing antibody. Applications Reported: This MQ1-17H12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MQ1-17H12 antibody has been pre-diluted and tested by flow cytometric analysis of activated normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor 532 is excited with the Green laser (532 nm) and emits at 561 nm. This cannot be used with the Yellow-Green laser (561 nm). We recommend using a 560/14 band pass filter.  Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 532 nm; Emission: 561 nm; Laser: Green Laser

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Spezifikation

Spezifikation

IL-2
5 μl/Test
Monoclonal
Liquid
RUO
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
3558
IgG2a κ
Affinity Chromatography
Antibody
MQ1-17H12
Flow Cytometry
Alexa Fluor 532
Rat
Human
P60568
IL2
Primary
4° C, store in dark, DO NOT FREEZE!
IL2
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen