missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-28A Mouse anti-Human, Clone: 3B4A1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60270-1-IG-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
IL28 is a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28B (IL28B), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).Spezifikation
IL-28A | |
Monoclonal | |
1.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IZJ0 | |
IFNL2 | |
IL-28A Fusion Protein Ag18525 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
3B4A1 | |
Unconjugated | |
IFNL2 | |
Cytokine Zcyto20, IFN lambda 2, IFNL2, IL 28A, IL28A, IL-28A, Interferon lambda 2, Interleukin 28A, ZCYTO20 | |
Mouse | |
Protein G | |
RUO | |
282616 | |
-20°C | |
Liquid |