missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-38/IL-1F10 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
N.V.
Spezifikation
Antigen | IL-38/IL-1F10 |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 μg/mL, ELISA |
Anwendungen | Western Blot, ELISA |
Klassifikation | Polyclonal |
Beschreibung
IL-38/IL-1F10 Polyclonal specifically detects IL-38/IL-1F10 in Human samples. It is validated for Western Blot, ELISA.Spezifikation
IL-38/IL-1F10 | |
Western Blot 0.1 μg/mL, ELISA | |
Polyclonal | |
Goat | |
Human | |
FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 | |
IL1F10 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Q8WWZ1 | |
84639 | |
E. coli-derived recombinant human IL-38/IL-1F10 Cys2-Trp152 Accession # Q8WWZ1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human IL-38/IL-1F10 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) IL-1ra and rhIL-1F5 is observed and less than 1% cross-reactivity with rhIL-1 alpha , rhIL-1 beta , rhIL-18, rhIL-1F6, rhIL-1F7, rhIL-1F8, and rhIL-1 F9 is observed. |
For Research Use Only