missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine IL-4 Antibody, R&D Systems™
Mouse Monoclonal Antibody
N.V.
Spezifikation
Antigen | IL-4 |
---|---|
Klon | 99605 |
Konzentration | LYOPH |
Verdünnung | Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Anwendungen | Western Blot, Flow Cytometry, CyTOF |
Beschreibung
IL-4 Monoclonal specifically detects IL-4 in Porcine samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Spezifikation
IL-4 | |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Porcine | |
Q04745 | |
3565 | |
E. coli-derived recombinant porcine IL-4 His25-Cys133 Accession # Q04745 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects porcine IL-4 in direct ELISAs and Western blots. Does not cross-react with recombinant human IL-13 or with recombinant IL-4 from human, canine, bovine, mouse, rat, cotton rat, or feline systems. |
99605 | |
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
IL4 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only