Learn More
IL1RL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553308
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: QAILTHSGKQ YEVLNGITVS ITERAGYGGS VPKIIYPKNH SIEVQLGTTL IVDCNVTDTK DNTNLRCWRV NNTLVDDYYD ESKRIREGVE THVSFREHNL YTVNITFLEV KMEDYGLPFM CHAG Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 72%.
IL1RL2 Polyclonal antibody specifically detects IL1RL2 in Human samples. It is validated for Immunohistochemistry.Spezifikation
IL1RL2 | |
Polyclonal | |
Unconjugated | |
IL1RL2 | |
AI481289; AW551444; IL-1 receptor-related protein 2; IL1RL2; IL1RRP2; IL-1Rrp2; IL1R-rp2; IL-36 receptor; IL-36R; interleukin 1 receptor like 2; interleukin 1 receptor-like 2; interleukin-1 receptor related protein 2; interleukin-1 receptor-like 2; Interleukin-1 receptor-related protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8808 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HB29 | |
IL1RL2 | |
Recombinant protein corresponding to Human IL1RL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only