Learn More
ILDR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553755
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LEYLEKELRN LNLAQPLPPD LKGRFGHPCS MLSSLGSEVV ERRIIHLPPL IRDLSSSRRT SDSLHQQWLT PIPSRPWDLR EGRSHHHYPD FHQELQDRGP KSWALERREL DPSWSGRHRS SRLNGSPIHW SDRDSLSDVP S Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 68%.
ILDR1 Polyclonal antibody specifically detects ILDR1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
ILDR1 | |
Polyclonal | |
Unconjugated | |
ILDR1 | |
AU041483; AU044638; DFNB42; ILDR1; ILDR1alpha; ILDR1alpha'; ILDR1beta; immunoglobulin like domain containing receptor 1; immunoglobulin-like domain containing receptor 1; immunoglobulin-like domain-containing receptor 1; immunoglobulin-like domain-containing receptor 1 alpha; immunoglobulin-like domain-containing receptor 1 beta; RGD1307792 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
286676 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86SU0 | |
ILDR1 | |
Recombinant protein corresponding to Human ILDR1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only