missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16205-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
IMP4 forms a ternary complex with IMP3.Spezifikation
IMP4 | |
Polyclonal | |
Unconjugated | |
IMP4 | |
BXDC4, IMP4, U3 snoRNP protein IMP4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27993, 316317, 92856 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5PQR5, Q8VHZ7, Q96G21 | |
Imp4 | |
IMP4 Fusion Protein Ag9175 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |