Description
Enable consistent results with Thermo Scientific™ Indiko™ System Electrodes. Specifically designed for electrolyte measurements, these ion selective electrodes (ISE) provide precise information about the electrolyte status, even with small sample sizes, thanks to direct measurement methods. The ready-to-use electrodes are an integral component of the Thermo Scientific™ Indiko™ analyzers, equipped with the ISE unit.
Direct measurement method, generally considered the most accurate ISE method, is used in Indiko™ analyzers.
- Simultaneous electrolyte measurement with photometric tests is possible in analyzers equipped with the ISE unit
- Electrode block relocates in its own dispensing arm
- Electrodes are ready to use: simply open the package and snap the potassium, sodium and chloride electrode into position
Specifications
Specifications
| Detectable Analytes | Potassium, Sodium, Chloride, Calcium, pH, Lithium |
| Sample Type | Plasma, Serum |
| Type | ISE Calibrator 1 |
| Quantity | 4 x 400 mL |