Learn More
Kartell SpA Industrial Tank
Product Code. 18852960
Change view
Click to view available options
Capacity (Metric):
2 L
Unit Size:
Pack of 64
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
18852960 2 L Pack of 64
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18852960 Supplier Kartell SpA Supplier No. 0178000
Verfügbar ab: 22-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Industrial tank with cover
This tank features a standardized industrial packaging design but could also be useful in everyday laboratory practice. Made of food grade HDPE, with a comfortable handle, and features a secure seal-cap following DIN recommendations.
Conforms to UN regulation on transportation of dangerous and toxic liquids and are suitable for road (ADR-RID), sea (IMO-IMDG) and air (IATA-ICAO) transport
industrial,tank,cover,hdpe,food grade
Specifications
Closure Size GL45
Type Tank with Cover
Material HDPE
Capacity (Metric) 2 L