missing translation for 'onlineSavingsMsg'
Learn More

INF2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16850015 Shop All Proteintech Products
Change view
Click to view available options
Menge:
20 μL
150 μL
Unit Size:
150 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
16850015 20 μL 20 Mikroliter
16840015 150 μL 150 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16850015 Supplier Proteintech Supplier No. 204661AP20UL

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene represents a member of the formin family of proteins. It is considered a diaphanous formin due to the presence of a diaphanous inhibitory domain located at the N-terminus of the encoded protein. Studies of a similar mouse protein indicate that the protein encoded by this locus may function in polymerization and depolymerization of actin filaments. Mutations at this locus have been associated with focal segmental glomerulosclerosis 5.

Specifications

Antigen INF2
Anwendungen Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konzentration 0.35 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen INF2
Gen-Zugriffsnummer Q0GNC1, Q27J81
Gen-Alias C14orf151, C14orf173, HBEBP2 binding protein C, INF2, Inverted formin 2, pp9484
Gensymbole INF2
Wirtsspezies Rabbit
Immunogen INF2 Fusion Protein Ag14302
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 64423, 70435
Zielspezies Mouse, Human
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.