missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ING4 Polyclonal specifically detects ING4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | ING4 |
| Anwendungen | Western Blot, ELISA |
| Klassifikation | Polyclonal |
| Konjugat | HRP |
| Verdünnung | Western Blot, ELISA |
| Zusammensetzung | PBS |
| Gen-Alias | candidate tumor suppressor p33 ING1 homolog, inhibitor of growth family, member 4, inhibitor of growth protein 4, MGC12557, my036, p29ING4brain my036 protein |
| Wirtsspezies | Goat |
| Immunogen | This affinity purified ING4 Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 150-175 of Human ING4 protein (Inhibitor of growth family, member 4). This sequence only shows homology to isoform 1 for ING4. (Uniprot: Q9UNL4) |
| Reinigungsverfahren | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?