missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA551331
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total ING5 protein.
ING5 Polyclonal antibody specifically detects ING5 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
ING5 | |
Polyclonal | |
Unconjugated | |
ING5 | |
1700001C14Rik; 1700027H23Rik; 1810018M11Rik; AI225768; ING5; inhibitor of growth family member 5; inhibitor of growth family, member 5; inhibitor of growth protein 5; p28ING5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84289 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8WYH8 | |
ING5 | |
Fusion protein of human ING5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |