missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inosin-5'-Monophosphat-Dinatriumsalsalzhydrat

Catalog Number 15464439
Click to view available options
Menge:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
352195-40-5
C10H11N4Na2O8P
392.17
MFCD00036201,MFCD00036201
AANLCWYVVNBGEE-IDIVVRGQSA-L
disodium 5'-inosinate
123134061
[(2R,3S,4R,5R)-3,4-Dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyldihydrogenphosphat;natrium
[Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1NC=NC2=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
352195-40-5
C10H11N4Na2O8P
392.17
MFCD00036201,MFCD00036201
AANLCWYVVNBGEE-IDIVVRGQSA-L
disodium 5'-inosinate
123134061
[(2R,3S,4R,5R)-3,4-Dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyldihydrogenphosphat;natrium
[Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1NC=NC2=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Inosine-5'-monophosphate disodium salt hydrate
Farbe Weiß
Menge 25 g
Formelmasse 392.17 (Anhydrous)
Physikalische Form Kristallpulver

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'