Learn More
INPP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557778
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: TSEKETIKAA LSRVCGDRIF GAAGAGYKSL CVVQGLVDIY IFSEDTTFKW DSCAAHAILR AMGGGIVDLK ECLERNPETG LDLPQLVY Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
INPP1 Polyclonal antibody specifically detects INPP1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
INPP1 | |
Polyclonal | |
Unconjugated | |
INPP1 | |
2300002C06Rik; AV137389; Inositol polyphosphate 1-phosphatase; inositol polyphosphate-1-phosphatase; Inpp1; IPP; IPPase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3628 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49441 | |
INPP1 | |
Recombinant protein corresponding to Human INPP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only