missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin alpha 9 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 19 publications
N.V.
Spezifikation
Antigen | Integrin alpha 9 |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 1-15 ug/mL, CyTOF-ready |
Anwendungen | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF |
Klassifikation | Polyclonal |
Beschreibung
Integrin alpha 9 Polyclonal specifically detects Integrin alpha 9 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Spezifikation
Integrin alpha 9 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 1-15 ug/mL, CyTOF-ready | |
Polyclonal | |
Goat | |
Mouse | |
ALPHA-RLC, integrin alpha-9, Integrin alpha-RLC, integrin, alpha 9, ITGA4L, ITGA9, RLC, RLC-alpha | |
ITGA9 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
RUO | |
NP_598482 | |
3680 | |
Mouse myeloma cell line NS0-derived recombinant mouse Integin alpha 9 Try31-Val979 Accession # NP_598482 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Integrin alpha 9 in direct ELISAs and Western blots. In direct ELISAs, less than 25% cross-reactivity with recombinant human Integrin alpha 9 is observed and less than 5% cross-reactivity with recombinant mouse Integrin alpha 3 and alpha 4 is observed. |
For Research Use Only