Learn More
INTS6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551599
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KYELEPSPLT QFILERKSPQ TCWQVYVSNS AKYSELGHPF GYLKASTALN CVNLFVMPYN YPVLLPLLDD LFKVHKAKPT LKWRQSFESY LKTMPPYYLG PLKKAVRMMG APNLIADSME YGLSYSVISY LKKLSQQAKI ES Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
INTS6 Polyclonal antibody specifically detects INTS6 in Human samples. It is validated for Immunohistochemistry.Spezifikation
INTS6 | |
Polyclonal | |
Unconjugated | |
INTS6 | |
2900075H24Rik; AI480962; Dbi1; DBI-1; Ddx26; DDX26A; DEAD box protein; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26; DICE1; DKFZP434B105; HDB; INT6; integrator complex subunit 6; INTS6; LRRGT00024; Notch2l; Notchl2; Protein DDX26; Protein deleted in cancer 1; RNA helicase HDB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26512 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UL03 | |
INTS6 | |
Recombinant protein corresponding to Human INTS6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only