missing translation for 'onlineSavingsMsg'
Learn More

INTU Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 24914-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16883425

  • 112.71 € / 20 Mikroliter

Verfügbar ab
17-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Plays a key role in ciliogenesis and embryonic development. Regulator of cilia formation by controlling the organization of the apical actin cytoskeleton and the positioning of the basal bodies at the apical cell surface, which in turn is essential for the normal orientation of elongating ciliary microtubules. Plays a key role in definition of cell polarity via its role in ciliogenesis but not via conversion extension. Has an indirect effect on hedgehog signaling
Spezifikation

Spezifikation

INTU
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q059U7, Q9ULD6
INT, INTU, KIAA1284, PDZD6, PDZK6
Rabbit
IgG
20 μL
-20°C
Primary
27152, 380614
Western Blot
Unconjugated
INTU
Liquid
Intu
INTU Fusion Protein Ag19313
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation