Learn More
Invitrogen™ E-Gel™ iBase™ USB Mini Cable
Enables uploading
Marke: Invitrogen™ G6300
Weitere Details : Netto-Gewicht : 0.00450kg
Beinhaltet: 1 mini USB cable
Beschreibung
E-Gel iBase USB Mini Cable enables uploading of new pre-set programs from the web site onto the E-Gel iBase Power System.
DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting, Proteins, Expression, Isolation and Analysis, Southern Blotting, Western Blotting
Spezifikation
USB Mini Cable | |
1 Cable |
For Research Use Only. Not for use in diagnostic procedures.