Learn More
(Iodomethyl)cyclopentane, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 27935-87-1 |
| Molecular Formula | C6H11I |
| MDL Number | MFCD03453190 |
| Quantity | 100 mg |
| SMILES | ICC1CCCC1 |
| Molecular Weight (g/mol) | 210.06 |
| Percent Purity | 98% |
| Chemical Name or Material | (Iodomethyl)cyclopentane |