Learn More
IPMK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558132
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DCFDGVLLEL RKYLPKYYGI WSPPTAPNDL YLKLEDVTHK FNKPCIMDVK IGQKSYDPFA SSEKIQQQVS KYPLMEEIGF LVLGMRVYHV HSD Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.
This gene encodes a member of the inositol phosphokinase family. The encoded protein has 3-kinase, 5-kinase and 6-kinase activities on phosphorylated inositol substrates. The encoded protein plays an important role in the biosynthesis of inositol 1, 3, 4, 5, 6-pentakisphosphate, and has a preferred 5-kinase activity. This gene may play a role in nuclear mRNA export. Pseudogenes of this gene are located on the long arm of chromosome 13 and the short arm of chromosome 19.Spezifikation
IPMK | |
Polyclonal | |
Unconjugated | |
IPMK | |
2410017C19Rik; AA408208; Impk; Inositol 1,3,4,6-tetrakisphosphate 5-kinase; inositol polyphosphate kinase 2; inositol polyphosphate multikinase; Ipk2; Ipmk; LOW QUALITY PROTEIN: inositol polyphosphate multikinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
253430 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFU5 | |
IPMK | |
Recombinant protein corresponding to Human IPMK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only