missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin 13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550432
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total IPO13 protein.
Importin 13 Polyclonal antibody specifically detects Importin 13 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
Importin 13 | |
Polyclonal | |
Unconjugated | |
IPO13 | |
Imp13; importin 13; importin-13; Ipo13; Kap13; karyopherin 13; karyopherin-13; KIAA0724; late gestation lung 2; late gestation lung 2 protein; LGL2; Ran binding protein 13; ran-binding protein 13; RanBP13; RP11-7O11.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9670 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O94829 | |
IPO13 | |
Fusion protein corresponding to residues near the C terminal of human importin 13 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |